4-(1-Adamantyl)-1,3-thiazol-2-amine

Supplier: Matrix Scientific

027038-500MG
101823-372EA 193.85 USD
101823-372
4-(1-Adamantyl)-1,3-thiazol-2-amine
4-(1-Adamantyl)-1,3-thiazol-2-amine

Hazard: Irritant

Formula: C₁₃H₁₈N₂S
MDL Number: MFCD00180700
CAS Number: 19735-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR