4,5,6,7-Tetrahydro[1,3]thiazolo[5,4-c]pyridin-2-amine dihydrobromide

Supplier: Matrix Scientific

027030-500MG
101823-356EA 300 USD
101823-356
4,5,6,7-Tetrahydro[1,3]thiazolo[5,4-c]pyridin-2-amine dihydrobromide
4,5,6,7-Tetrahydro[1,3]thiazolo[5,4-c]pyridin-2-amine dihydrobromide

Hazard: Irritant

Formula: C₆H₁₁Br₂N₃S
MDL Number: MFCD11506406
CAS Number: 1184964-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR