N1-[4-(Diethylamino)benzyl]-N3-methyl-1,3-propanediamine

Supplier: Matrix Scientific

027022-500MG
101823-338EA 306.15 USD
101823-338
N1-[4-(Diethylamino)benzyl]-N3-methyl-1,3-propanediamine
N1-[4-(Diethylamino)benzyl]-N3-methyl-1,3-propanediamine

Hazard: Irritant

Formula: C₁₅H₂₇N₃
MDL Number: MFCD07408088
CAS Number: 892567-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR