N1-Methyl-N3-(3-pyridinylmethyl)-1,3-propanediamine

Supplier: Matrix Scientific

027018-500MG
101823-330EA 306.15 USD
101823-330
N1-Methyl-N3-(3-pyridinylmethyl)-1,3-propanediamine
N1-Methyl-N3-(3-pyridinylmethyl)-1,3-propanediamine

Hazard: Irritant

Formula: C₁₀H₁₇N₃
MDL Number: MFCD07408154
CAS Number: 892566-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR