N-Ethyl-3-[(3-pyridinylmethyl)amino]propanamide

Supplier: Matrix Scientific

027017-500MG
101823-328EA 306.15 USD
101823-328
N-Ethyl-3-[(3-pyridinylmethyl)amino]propanamide
N-Ethyl-3-[(3-pyridinylmethyl)amino]propanamide

Hazard: Irritant

Formula: C₁₁H₁₇N₃O
MDL Number: MFCD10687391
CAS Number: 1040693-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR