N1,N1-Diethyl-N3-methyl-1,3-propanediamine

Supplier: Matrix Scientific

027015-500MG
101823-324EA 306.15 USD
101823-324
N1,N1-Diethyl-N3-methyl-1,3-propanediamine
N1,N1-Diethyl-N3-methyl-1,3-propanediamine

Hazard: Irritant/Stored under Argon

Formula: C₈H₂₀N₂
MDL Number: MFCD00026748
CAS Number: 5459-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR