3-[(2-Chlorobenzyl)amino]-N-ethylpropanamide

Supplier: Matrix Scientific

027014-500MG
101823-322EA 306.15 USD
101823-322
3-[(2-Chlorobenzyl)amino]-N-ethylpropanamide
3-[(2-Chlorobenzyl)amino]-N-ethylpropanamide

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD10687389
CAS Number: 1040693-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR