3-[(2-Chlorobenzyl)amino]-N-methylpropanamide
Supplier: Matrix Scientific
027013-500MG
101823-320EA
306.15
USD
101823-320
3-[(2-Chlorobenzyl)amino]-N-methylpropanamide
3-[(2-Chlorobenzyl)amino]-N-methylpropanamide
Hazard: Irritant
Formula:
C₁₁H₁₅ClN₂O |
MDL Number:
MFCD10687388 CAS Number: 1040693-35-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...