N1-(3-Methoxypropyl)-N3-methyl-1,3-propanediamine

Supplier: Matrix Scientific

027011-500MG
101823-316EA 306.15 USD
101823-316
N1-(3-Methoxypropyl)-N3-methyl-1,3-propanediamine
N1-(3-Methoxypropyl)-N3-methyl-1,3-propanediamine

Hazard: Irritant

Formula: C₈H₂₀N₂O
MDL Number: MFCD10687386
CAS Number: 1040693-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR