N1-(sec-Butyl)-N3-ethyl-1,3-propanediamine

Supplier: Matrix Scientific

027008-500MG
101823-310EA 306.15 USD
101823-310
N1-(sec-Butyl)-N3-ethyl-1,3-propanediamine
N1-(sec-Butyl)-N3-ethyl-1,3-propanediamine

Hazard: Irritant

Formula: C₉H₂₂N₂
MDL Number: MFCD10687383
CAS Number: 1040692-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR