N1-(sec-Butyl)-N3-methyl-1,3-propanediamine

Supplier: Matrix Scientific

027007-500MG
101823-308EA 306.15 USD
101823-308
N1-(sec-Butyl)-N3-methyl-1,3-propanediamine
N1-(sec-Butyl)-N3-methyl-1,3-propanediamine

Hazard: Irritant

Formula: C₈H₂₀N₂
MDL Number: MFCD10687382
CAS Number: 1040692-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR