N-Ethyl-3-[(3-hydroxypropyl)amino]propanamide

Supplier: Matrix Scientific

027001-500MG
101823-296EA 306.15 USD
101823-296
N-Ethyl-3-[(3-hydroxypropyl)amino]propanamide
N-Ethyl-3-[(3-hydroxypropyl)amino]propanamide

Hazard: Irritant

Formula: C₈H₁₈N₂O₂
MDL Number: MFCD10687377
CAS Number: 1040692-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR