N1,N3-Diethyl-1,3-propanediamine

Supplier: Matrix Scientific

026995-500MG
101823-284EA 306.15 USD
101823-284
N1,N3-Diethyl-1,3-propanediamine
N1,N3-Diethyl-1,3-propanediamine

Hazard: Irritant

Formula: C₇H₁₈N₂
MDL Number: MFCD00009036
CAS Number: 10061-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR