N-Ethyl-3-(ethylamino)propanamide

Supplier: Matrix Scientific

026994-500MG
101823-282EA 306.15 USD
101823-282
N-Ethyl-3-(ethylamino)propanamide
N-Ethyl-3-(ethylamino)propanamide

Hazard: Irritant

Formula: C₇H₁₆N₂O
MDL Number: MFCD10687372
CAS Number: 31035-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR