3-(4-Benzhydryl-1-piperazinyl)-N-methyl-1-propanamine

Supplier: Matrix Scientific

026991-500MG
101823-276EA 306.15 USD
101823-276
3-(4-Benzhydryl-1-piperazinyl)-N-methyl-1-propanamine
3-(4-Benzhydryl-1-piperazinyl)-N-methyl-1-propanamine

Hazard: Irritant

Formula: C₂₁H₂₉N₃
MDL Number: MFCD10687370
CAS Number: 1040692-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR