N1-Ethyl-N3-propyl-1,3-propanediamine

Supplier: Matrix Scientific

026984-500MG
101823-262EA 306.15 USD
101823-262
N1-Ethyl-N3-propyl-1,3-propanediamine
N1-Ethyl-N3-propyl-1,3-propanediamine

Hazard: Irritant

Formula: C₈H₂₀N₂
MDL Number: MFCD10687363
CAS Number: 1040691-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR