N1-Methyl-N3-propyl-1,3-propanediamine

Supplier: Matrix Scientific

026983-500MG
101823-260EA 306.15 USD
101823-260
N1-Methyl-N3-propyl-1,3-propanediamine
N1-Methyl-N3-propyl-1,3-propanediamine

Hazard: Irritant

Formula: C₇H₁₈N₂
MDL Number: MFCD10687362
CAS Number: 50910-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR