N1-Benzyl-N3-ethyl-N1-methyl-1,3-propanediamine

Supplier: Matrix Scientific

026982-500MG
101823-258EA 306.15 USD
101823-258
N1-Benzyl-N3-ethyl-N1-methyl-1,3-propanediamine
N1-Benzyl-N3-ethyl-N1-methyl-1,3-propanediamine

Hazard: Irritant

Formula: C₁₃H₂₂N₂
MDL Number: MFCD10687361
CAS Number: 98902-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR