N1-(2-Furylmethyl)-N3-methyl-1,3-propanediamine
Supplier: Matrix Scientific
026978-500MG
101823-250EA
306.15
USD
101823-250
N1-(2-Furylmethyl)-N3-methyl-1,3-propanediamine
N1-(2-Furylmethyl)-N3-methyl-1,3-propanediamine
Hazard: Irritant
Formula:
C₉H₁₆N₂O |
MDL Number:
MFCD07410493 CAS Number: 892582-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...