N-Ethyl-3-[(2-furylmethyl)amino]propanamide

Supplier: Matrix Scientific

026977-500MG
101823-248EA 306.15 USD
101823-248
N-Ethyl-3-[(2-furylmethyl)amino]propanamide
N-Ethyl-3-[(2-furylmethyl)amino]propanamide

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₂
MDL Number: MFCD10687358
CAS Number: 1040691-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR