N1-Ethyl-N3,N3-dipropyl-1,3-propanediamine

Supplier: Matrix Scientific

026975-500MG
101823-244EA 306.15 USD
101823-244
N1-Ethyl-N3,N3-dipropyl-1,3-propanediamine
N1-Ethyl-N3,N3-dipropyl-1,3-propanediamine

Hazard: Irritant

Formula: C₁₁H₂₆N₂
MDL Number: MFCD10687356
CAS Number: 1040691-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR