N1-Benzyl-N1,N3-dimethyl-1,3-propanediamine

Supplier: Matrix Scientific

026971-500MG
101823-236EA 306.15 USD
101823-236
N1-Benzyl-N1,N3-dimethyl-1,3-propanediamine
N1-Benzyl-N1,N3-dimethyl-1,3-propanediamine

Hazard: Irritant

Formula: C₁₂H₂₀N₂
MDL Number: MFCD10687352
CAS Number: 60630-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR