N1-Ethyl-N3-isopropyl-1,3-propanediamine

Supplier: Matrix Scientific

026970-500MG
101823-234EA 306.15 USD
101823-234
N1-Ethyl-N3-isopropyl-1,3-propanediamine
N1-Ethyl-N3-isopropyl-1,3-propanediamine

Hazard: Irritant

Formula: C₈H₂₀N₂
MDL Number: MFCD10687351
CAS Number: 78706-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR