N1-Isopropyl-N3-methyl-1,3-propanediamine

Supplier: Matrix Scientific

026969-500MG
101823-232EA 384.62 USD
101823-232
N1-Isopropyl-N3-methyl-1,3-propanediamine
N1-Isopropyl-N3-methyl-1,3-propanediamine

Hazard: Irritant

Formula: C₇H₁₈N₂
MDL Number: MFCD10687350
CAS Number: 15937-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR