N-Ethyl-3-(isopropylamino)propanamide

Supplier: Matrix Scientific

026968-500MG
101823-230EA 306.15 USD
101823-230
N-Ethyl-3-(isopropylamino)propanamide
N-Ethyl-3-(isopropylamino)propanamide

Hazard: Irritant

Formula: C₈H₁₈N₂O
MDL Number: MFCD10687349
CAS Number: 1040691-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR