N1-Cyclohexyl-N3-ethyl-1,3-propanediamine

Supplier: Matrix Scientific

026964-500MG
101823-222EA 306.15 USD
101823-222
N1-Cyclohexyl-N3-ethyl-1,3-propanediamine
N1-Cyclohexyl-N3-ethyl-1,3-propanediamine

Hazard: Irritant

Formula: C₁₁H₂₄N₂
MDL Number: MFCD10687346
CAS Number: 1026885-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR