N1-Methyl-N3-(tetrahydro-2-furanylmethyl)-1,3-propanediamine

Supplier: Matrix Scientific

026956-500MG
101823-206EA 306.15 USD
101823-206
N1-Methyl-N3-(tetrahydro-2-furanylmethyl)-1,3-propanediamine
N1-Methyl-N3-(tetrahydro-2-furanylmethyl)-1,3-propanediamine

Hazard: Irritant

Formula: C₉H₂₀N₂O
MDL Number: MFCD10687338
CAS Number: 1040690-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR