N-Ethyl-3-(4-methyl-1-piperidinyl)-1-propanamine

Supplier: Matrix Scientific

026951-500MG
101823-196EA 306.15 USD
101823-196
N-Ethyl-3-(4-methyl-1-piperidinyl)-1-propanamine
N-Ethyl-3-(4-methyl-1-piperidinyl)-1-propanamine

Hazard: Irritant

Formula: C₁₁H₂₄N₂
MDL Number: MFCD10687335
CAS Number: 1040690-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR