N1-Hexadecyl-N3-methyl-1,3-propanediamine

Supplier: Matrix Scientific

026949-500MG
101823-192EA 306.15 USD
101823-192
N1-Hexadecyl-N3-methyl-1,3-propanediamine
N1-Hexadecyl-N3-methyl-1,3-propanediamine

Hazard: Irritant

Formula: C₂₀H₄₄N₂
MDL Number: MFCD10687333
CAS Number: 1040690-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR