N1-Ethyl-N3-heptyl-1,3-propanediamine

Supplier: Matrix Scientific

026948-500MG
101823-190EA 306.15 USD
101823-190
N1-Ethyl-N3-heptyl-1,3-propanediamine
N1-Ethyl-N3-heptyl-1,3-propanediamine

Hazard: Irritant

Formula: C₁₂H₂₈N₂
MDL Number: MFCD10687332
CAS Number: 1040690-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR