3-(1-Azepanyl)-N-ethyl-1-propanamine

Supplier: Matrix Scientific

026944-500MG
101823-182EA 306.15 USD
101823-182
3-(1-Azepanyl)-N-ethyl-1-propanamine
3-(1-Azepanyl)-N-ethyl-1-propanamine

Hazard: Irritant

Formula: C₁₁H₂₄N₂
MDL Number: MFCD10687328
CAS Number: 1040690-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR