N1-Ethyl-N3-(1-phenylethyl)-1,3-propanediamine

Supplier: Matrix Scientific

026942-500MG
101823-178EA 306.15 USD
101823-178
N1-Ethyl-N3-(1-phenylethyl)-1,3-propanediamine
N1-Ethyl-N3-(1-phenylethyl)-1,3-propanediamine

Hazard: Irritant

Formula: C₁₃H₂₂N₂
MDL Number: MFCD10687327
CAS Number: 1040690-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR