N1-Methyl-N3-(1-phenylethyl)-1,3-propanediamine

Supplier: Matrix Scientific

026941-500MG
101823-176EA 306.15 USD
101823-176
N1-Methyl-N3-(1-phenylethyl)-1,3-propanediamine
N1-Methyl-N3-(1-phenylethyl)-1,3-propanediamine

Hazard: Irritant

Formula: C₁₂H₂₀N₂
MDL Number: MFCD10687326
CAS Number: 1009511-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR