N-Ethyl-3-[(1-phenylethyl)amino]propanamide

Supplier: Matrix Scientific

026940-500MG
101823-174EA 306.15 USD
101823-174
N-Ethyl-3-[(1-phenylethyl)amino]propanamide
N-Ethyl-3-[(1-phenylethyl)amino]propanamide

Hazard: Irritant

Formula: C₁₃H₂₀N₂O
MDL Number: MFCD10687325
CAS Number: 1040690-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR