N-Ethyl-3-(hexadecylamino)propanamide

Supplier: Matrix Scientific

026938-500MG
101823-170EA 306.15 USD
101823-170
N-Ethyl-3-(hexadecylamino)propanamide
N-Ethyl-3-(hexadecylamino)propanamide

Hazard: Irritant

Formula: C₂₁H₄₄N₂O
MDL Number: MFCD10687323
CAS Number: 1040690-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR