3-(tert-Butylamino)-N-methylpropanamide

Supplier: Matrix Scientific

026933-500MG
101823-160EA 306.15 USD
101823-160
3-(tert-Butylamino)-N-methylpropanamide
3-(tert-Butylamino)-N-methylpropanamide

Hazard: Irritant

Formula: C₈H₁₈N₂O
MDL Number: MFCD10687319
CAS Number: 1040689-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR