N1-Benzyl-N3-ethyl-1,3-propanediamine

Supplier: Matrix Scientific

026932-500MG
101823-158EA 306.15 USD
101823-158
N1-Benzyl-N3-ethyl-1,3-propanediamine
N1-Benzyl-N3-ethyl-1,3-propanediamine

Hazard: Irritant

Formula: C₁₂H₂₀N₂
MDL Number: MFCD09693117
CAS Number: 57857-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR