Methyl-3-[(2-thienylmethyl)amino]propanoate

Supplier: Matrix Scientific

026925-500MG
101823-144EA 306.15 USD
101823-144
Methyl-3-[(2-thienylmethyl)amino]propanoate
Methyl-3-[(2-thienylmethyl)amino]propanoate

Hazard: Irritant

Formula: C₉H₁₃NO₂S
MDL Number: MFCD10687315
CAS Number: 146033-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR