3-(Cyclopentylamino)propanamide

Supplier: Matrix Scientific

026924-500MG
101823-142EA 306.15 USD
101823-142
3-(Cyclopentylamino)propanamide
3-(Cyclopentylamino)propanamide

Hazard: Irritant

Formula: C₈H₁₆N₂O
MDL Number: MFCD10687314
CAS Number: 192811-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR