Ethyl-3-(cyclopentylamino)propanoate

Supplier: Matrix Scientific

026923-500MG
101823-140EA 306.15 USD
101823-140
Ethyl-3-(cyclopentylamino)propanoate
Ethyl-3-(cyclopentylamino)propanoate

Hazard: Irritant

Formula: C₁₀H₁₉NO₂
MDL Number: MFCD10687313
CAS Number: 928788-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR