3-(Methylamino)propanamide hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-(Methylamino)propanamide

026919-500MG
101823-132EA 306.15 USD
101823-132
3-(Methylamino)propanamide hydrochloride
3-(Methylamino)propanamide hydrochloride

Hazard: Irritant

Formula: C₄H₁₁ClN₂O
MW: 138.6 g/mol
MDL Number: MFCD11226474
CAS Number: 4874-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR