3-(4-Benzhydryl-1-piperazinyl)-1-propanamine

Supplier: Matrix Scientific

026913-500MG
101823-120EA 306.15 USD
101823-120
3-(4-Benzhydryl-1-piperazinyl)-1-propanamine
3-(4-Benzhydryl-1-piperazinyl)-1-propanamine

Hazard: Irritant

Formula: C₂₀H₂₇N₃
MDL Number: MFCD10687310
CAS Number: 50971-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR