3-[(2-Methoxyethyl)amino]propanohydrazide

Supplier: Matrix Scientific

026912-500MG
101823-118EA 306.15 USD
101823-118
3-[(2-Methoxyethyl)amino]propanohydrazide
3-[(2-Methoxyethyl)amino]propanohydrazide

Hazard: Irritant

Formula: C₆H₁₅N₃O₂
MDL Number: MFCD10687309
CAS Number: 1040689-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR