N1-(2-Methoxyethyl)-1,3-propanediamine

Supplier: Matrix Scientific

026911-500MG
101823-116EA 306.15 USD
101823-116
N1-(2-Methoxyethyl)-1,3-propanediamine
N1-(2-Methoxyethyl)-1,3-propanediamine

Hazard: Irritant

Formula: C₆H₁₆N₂O
MDL Number: MFCD10687308
CAS Number: 187150-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR