Ethyl-3-[(2-methoxyethyl)amino]propanoate

Supplier: Matrix Scientific

026908-500MG
101823-110EA 306.15 USD
101823-110
Ethyl-3-[(2-methoxyethyl)amino]propanoate
Ethyl-3-[(2-methoxyethyl)amino]propanoate

Hazard: Irritant

Formula: C₈H₁₇NO₃
MDL Number: MFCD01476077
CAS Number: 66092-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR