3-(Allylamino)propanenitrile

Supplier: Matrix Scientific

026904-500MG
101823-102EA 306.15 USD
101823-102
3-(Allylamino)propanenitrile
3-(Allylamino)propanenitrile

Hazard: Irritant

Formula: C₆H₁₀N₂
MDL Number: MFCD10687302
CAS Number: 34508-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR