3-(Allylamino)propanamide

Supplier: Matrix Scientific

026903-500MG
101823-100EA 306.15 USD
101823-100
3-(Allylamino)propanamide
3-(Allylamino)propanamide

Hazard: Irritant

Formula: C₆H₁₂N₂O
MDL Number: MFCD10687301
CAS Number: 24626-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR