Ethyl-3-(allylamino)propanoate

Supplier: Matrix Scientific

026902-500MG
101823-098EA 306.15 USD
101823-098
Ethyl-3-(allylamino)propanoate
Ethyl-3-(allylamino)propanoate

Hazard: Irritant

Formula: C₈H₁₅NO₂
MDL Number: MFCD10687300
CAS Number: 44981-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR