N-Propyl-1,3-propanediamine

Supplier: Matrix Scientific

026899-500MG
101823-092EA 228.85 USD
101823-092
N-Propyl-1,3-propanediamine
N-Propyl-1,3-propanediamine

Hazard: Irritant

Formula: C₆H₁₆N₂
UN: 2734
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR