Ethyl-3-(propylamino)propanoate

Supplier: Matrix Scientific

026896-500MG
101823-088EA 306.15 USD
101823-088
Ethyl-3-(propylamino)propanoate
Ethyl-3-(propylamino)propanoate

Hazard: Irritant

Formula: C₈H₁₇NO₂
MDL Number: MFCD01475851
CAS Number: 3186-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR